Gene Transcripts
- Gene Symbol
- Srpk1
- Entrez Gene ID
- 20815
- Gene Name
- serine/arginine-rich protein specific kinase 1
- Gene Aliases
- AU017960
- Chromosome Location
- Chr. 17: 28589590 - 28622636 on Build GRCm38
- UniGene ID
- Mm.15252
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016795.3 |
NP_058075.2
|
3, 4 |
246 |
|
GenBank mRNA |
AB012290.1 |
- |
2, 3 |
204 |
|
AJ224115.1 |
- |
3, 4 |
240 |
|
AK017187.1 |
- |
3, 4 |
243 |
|
AK141271.1 |
- |
3, 4 |
313 |
|
AK151152.1 |
- |
3, 4 |
309 |
|
AK161346.1 |
- |
3, 4 |
224 |
|
AK161388.1 |
- |
3, 4 |
225 |
|
BC005707.1 |
- |
3, 4 |
215 |
3157835 |
BC050761.1 |
- |
3, 4 |
246 |
6771151 |
View Full Product Details