Gene Transcripts

Gene Symbol
Hmgn5
Entrez Gene ID
50887
Gene Name
high-mobility group nucleosome binding domain 5
Gene Aliases
GARP45, NBP-45, Nsbp1
Chromosome Location
Chr. X: 109004537 - 109013380 on Build GRCm38
UniGene ID
Mm.298443
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016710.2 NP_057919.2 6 407
GenBank mRNA AB018374.2 - 6 362
AF213454.1 - 6 387
AK013748.1 - 5 276
AK131996.1 - 6 377
AK146755.1 - 6 383
BC021626.1 - 6 359 4219308
BC083087.1 - 6 369 6518798

View Full Product Details