Gene Transcripts

Gene Symbol
Tcirg1
Entrez Gene ID
27060
Gene Name
T cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 protein A3
Gene Aliases
ATP6N1C, ATP6a3, Atp6i, OC-116, OPTB1, Stv1, TIRC7, Vph1, oc
Chromosome Location
Chr. 19: 3896050 - 3907133 on Build GRCm38
UniGene ID
Mm.271689
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136091.2 NP_001129563.1 5 566
NM_001167784.1 NP_001161256.1 5 911
NM_016921.3 NP_058617.3 5 607
GenBank mRNA AB022322.1 - 5 473
AF218253.1 - 5 545
AK155140.1 - 5 912
BC006761.1 - 5 607 3257437
BC085234.1 - 5 467 5322429

View Full Product Details