Gene Transcripts

Gene Symbol
Lyz2
Entrez Gene ID
17105
Gene Name
lysozyme 2
Gene Aliases
AI326280, Lys, Lysm, Lyzs, Lzm, Lzm-s1, Lzp
Chromosome Location
Chr. 10: 117277541 - 117282272 on Build GRCm38
UniGene ID
Mm.45436
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017372.3 NP_059068.1 2 237
GenBank mRNA AK148516.1 - 2 238
AK150998.1 - 2 240
AK153244.1 - 2 240
AK153475.1 - 2 238
AK159276.1 - 2 287
AK159640.1 - 2 234
BC002069.1 - 2 229 3593360
BC054463.1 - 2 229 6514977

View Full Product Details