Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001113517.1 | NP_001106989.1 | 6 | 873 | |
NM_001113518.1 | NP_001106990.1 | 5 | 662 | ||
NM_017402.4 | NP_059098.2 | 5 | 662 | ||
GenBank mRNA | AF247654.1 | - | 5 | 558 | |
AF247655.1 | - | 3 | 279 | ||
AF343877.1 | - | 5 | 542 | ||
AK052545.1 | - | 5 | 633 | ||
AK129064.1 | - | 6 | 809 | ||
AK139156.1 | - | 6 | 802 | ||
AY220301.1 | - | 5 | 627 | ||
BC044838.1 | - | 5 | 647 | 3499604 | |
U96634.1 | - | 3 | 352 |