Gene Transcripts

Gene Symbol
Slc1a4
Entrez Gene ID
55963
Gene Name
solute carrier family 1 (glutamate/neutral amino acid transporter), member 4
Gene Aliases
ASCT-1, ASCT1, AW045657, SATT
Chromosome Location
Chr. 11: 20302180 - 20332713 on Build GRCm38
UniGene ID
Mm.6379
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018861.3 NP_061349.3 8 2358
GenBank mRNA AK044280.1 - 8 2329
AK136900.1 - 8 2358
AK139248.1 - 8 2357
AK140866.1 - 8 2359
AK148075.1 - 8 2359
BC043483.1 - 8 2316 5003744
BC052733.1 - 8 2232 6816689
U75215.1 - 8 2225

View Full Product Details