Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019447.2 | NP_062320.2 | 5, 6 | 628 | |
GenBank mRNA | AF099017.1 | - | 5, 6 | 588 | |
AF224724.1 | - | 5, 6 | 588 | ||
AK007987.1 | - | 5, 6 | 626 | ||
AK075718.1 | - | 5, 6 | 537 | ||
AK144554.1 | - | 5, 6 | 600 | ||
AK149522.1 | - | 5, 6 | 592 | ||
AK169989.1 | - | 5, 6 | 624 | ||
BC019376.1 | - | 5, 6 | 593 | 5134851 |