Gene Transcripts
- Gene Symbol
- Myo10
- Entrez Gene ID
- 17909
- Gene Name
- myosin X
- Gene Aliases
- AW048724, D15Ertd600e, mKIAA0799
- Chromosome Location
- Chr. 15: 25622550 - 25813673 on Build GRCm38
- UniGene ID
- Mm.60590
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AJ249706.2 |
- |
6, 7 |
804 |
|
View Full Product Details