Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019575.4 | NP_062521.1 | 7 | 723 | |
GenBank mRNA | AF224721.1 | - | 7 | 698 | |
AF295102.1 | - | 7 | 716 | ||
AK019112.1 | - | 7 | 727 | ||
AK078226.1 | - | 7 | 715 | ||
AK154094.1 | - | 7 | 714 | ||
AK160755.1 | - | 7 | 714 | ||
AK168554.1 | - | 7 | 709 | ||
BC018215.1 | - | 7 | 692 | 3965223 | |
BC054104.2 | - | 7 | 718 | 30021104 |