Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004982.3 | NP_004973.1 | 3 | 2354 | |
XM_005253358.4 | XP_005253415.1 | 4 | 2323 | ||
XM_017019283.1 | XP_016874772.1 | 4 | 2258 | ||
XM_017019284.1 | XP_016874773.1 | 4 | 2329 |