Gene Transcripts

Gene Symbol
Mlst8
Entrez Gene ID
56716
Gene Name
MTOR associated protein, LST8 homolog (S. cerevisiae)
Gene Aliases
0610033N12Rik, AA409454, AI505104, AI851821, Gbl
Chromosome Location
Chr. 17: 24473550 - 24479114 on Build GRCm38
UniGene ID
Mm.289516
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252463.1 NP_001239392.1 2, 3 233
NM_001252464.1 NP_001239393.1 2, 3 208
NM_019988.5 NP_064372.2 2, 3 228
GenBank mRNA AF237676.1 - 2, 3 159
AK002751.1 - 2, 3 194
AK038515.1 - 1, 2 726
AK040100.1 - 2, 3 229
AK048102.1 - 2, 3 192
AK077680.1 - 2, 3 199
AK083731.1 - 2, 3 169
AK088951.1 - 2, 3 201
BC015279.1 - 2, 3 187 4222934
CF750475.1 - 2, 3 197

View Full Product Details