Gene Transcripts

Gene Symbol
Vmn2r1
Entrez Gene ID
56544
Gene Name
vomeronasal 2, receptor 1
Gene Aliases
Casrl1, EG56544, V2r83
Chromosome Location
Chr. 3: 64079774 - 64220847 on Build GRCm38
UniGene ID
Mm.57069
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019918.2 NP_064302.2 1 168
GenBank mRNA AB267725.1 - 1 168
AF053986.1 - 1 168

View Full Product Details