Gene Transcripts
- Gene Symbol
- Rbms1
- Entrez Gene ID
- 56878
- Gene Name
- RNA binding motif, single stranded interacting protein 1
- Gene Aliases
- 2600014B10Rik, AI255215, MSSP-1, MSSP-2, MSSP-3, YC1
- Chromosome Location
- Chr. 2: 60750193 - 60963450 on Build GRCm38
- UniGene ID
- Mm.259667
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001141931.1 |
NP_001135403.1
|
3, 4 |
564 |
|
NM_001141932.1 |
NP_001135404.1
|
3, 4 |
564 |
|
NM_020296.2 |
NP_064692.2
|
3, 4 |
757 |
|
GenBank mRNA |
AB026569.1 |
- |
3, 4 |
480 |
|
AK133054.1 |
- |
3, 4 |
285 |
|
AK135280.1 |
- |
3, 4 |
444 |
|
AK145897.1 |
- |
3, 4 |
566 |
|
AK147109.1 |
- |
3, 4 |
758 |
|
AK161102.1 |
- |
3, 4 |
514 |
|
AK164307.1 |
- |
3, 4 |
558 |
|
BC016501.1 |
- |
3, 4 |
745 |
4527742 |
BC057866.1 |
- |
3, 4 |
596 |
5327143 |
View Full Product Details