Gene Transcripts
- Gene Symbol
- Tk2
- Entrez Gene ID
- 57813
- Gene Name
- thymidine kinase 2, mitochondrial
- Gene Aliases
- AU024611, mt-TK
- Chromosome Location
- Chr. 8: 104226691 - 104248574 on Build GRCm38
- UniGene ID
- Mm.183110
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021028.3 |
NP_066356.3
|
2, 3 |
189 |
|
GenBank mRNA |
AF105217.1 |
- |
2, 3 |
164 |
|
AJ249341.2 |
- |
2, 3 |
167 |
|
AK041651.1 |
- |
2, 3 |
181 |
|
AK051286.1 |
- |
2, 3 |
183 |
|
AK087557.1 |
- |
2, 3 |
190 |
|
AK154160.1 |
- |
2, 3 |
183 |
|
AK210286.1 |
- |
1, 2 |
34 |
|
BC019982.1 |
- |
2, 3 |
181 |
4159742 |
BY083126.1 |
- |
2, 3 |
184 |
|
View Full Product Details