Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021053.4 | NP_066395.3 | 1 | 278 | |
GenBank mRNA | AF148145.1 | - | 1 | 250 | |
AK014784.1 | - | 1 | 265 | ||
AK017363.1 | - | 1 | 278 | ||
AK040063.1 | - | 279 | |||
BC140962.1 | - | 1 | 211 | 9055574 |