Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001167730.1 | NP_001161202.1 | 5 | 417 | |
NM_021385.2 | NP_067360.2 | 5 | 417 | ||
GenBank mRNA | AF205278.1 | - | 5 | 341 | |
AK012795.1 | - | 5 | 418 | ||
AK028505.1 | - | 5 | 337 | ||
AK030043.1 | - | 5 | 418 | ||
AK048975.1 | - | 5 | 392 | ||
BC011120.1 | - | 5 | 380 | 4014013 |