Gene Transcripts
- Gene Symbol
- Ttyh1
- Entrez Gene ID
- 57776
- Gene Name
- tweety homolog 1 (Drosophila)
- Gene Aliases
- 4930459B04Rik, 6330408P11Rik, TTY1, mTTY1, tty
- Chromosome Location
- Chr. 7: 4119527 - 4136241 on Build GRCm38
- UniGene ID
- Mm.29729
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB162929.1 |
- |
10, 11 |
1139 |
|
AF190699.1 |
- |
10, 11 |
1211 |
|
AF190991.1 |
- |
10, 11 |
1211 |
|
AK015486.1 |
- |
9, 10 |
862 |
|
AK018148.1 |
- |
10, 11 |
1203 |
|
AK031550.1 |
- |
10, 11 |
1239 |
|
AK042758.1 |
- |
10, 11 |
992 |
|
AK043998.1 |
- |
10, 11 |
1259 |
|
AK048413.1 |
- |
10, 11 |
1210 |
|
AK049903.1 |
- |
9, 10 |
945 |
|
AK081981.1 |
- |
10, 11 |
1213 |
|
AK083126.1 |
- |
9, 10 |
1336 |
|
BC046310.1 |
- |
10, 11 |
1197 |
6307420 |
BC065694.1 |
- |
10, 11 |
1203 |
6813365 |
View Full Product Details