Gene Transcripts

Gene Symbol
Pmaip1
Entrez Gene ID
58801
Gene Name
phorbol-12-myristate-13-acetate-induced protein 1
Gene Aliases
Noxa
Chromosome Location
Chr. 18: 66458604 - 66465558 on Build GRCm38
UniGene ID
Mm.271878
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021451.2 NP_067426.1 3 381
GenBank mRNA AB041230.1 - 3 381
AK043856.1 - 3 412
AK088556.1 - 3 391
AK143990.1 - 3 453
AK169914.1 - 3 425
BC050821.1 - 3 368 6517820

View Full Product Details