Gene Transcripts

Gene Symbol
Midn
Entrez Gene ID
59090
Gene Name
midnolin
Gene Aliases
3000003C15Rik
Chromosome Location
Chr. 10: 80148227 - 80158368 on Build GRCm38
UniGene ID
Mm.143813
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB036882.1 - 5 888
AK154535.1 - 4 832
BC034719.1 - 5 858 3710931

View Full Product Details