Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022017.3 | NP_071300.2 | 9 | 1680 | |
GenBank mRNA | AB021875.2 | - | 8 | 1607 | |
AF208026.1 | - | 9 | 1648 | ||
AF263522.1 | - | 8 | 1638 | ||
AF279672.1 | - | 9 | 1664 | ||
AJ296078.1 | - | 8 | 1599 | ||
AK083223.1 | - | 9 | 1651 | ||
AK158652.1 | - | 5 | 1533 | ||
BC127052.1 | - | 9 | 1645 | 40109678 |