Gene Transcripts
- Gene Symbol
- Irak1bp1
- Entrez Gene ID
- 65099
- Gene Name
- interleukin-1 receptor-associated kinase 1 binding protein 1
- Gene Aliases
- 4921528N06Rik, AI851240, Aabp3, Aip70, Simpl
- Chromosome Location
- Chr. 9: 82829806 - 82847688 on Build GRCm38
- UniGene ID
- Mm.41415
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022986.4 |
NP_075362.3
|
1, 2 |
542 |
|
GenBank mRNA |
AF093135.1 |
- |
1, 2 |
325 |
|
AJ242722.1 |
- |
1, 2 |
167 |
|
AK013036.2 |
- |
1, 2 |
347 |
|
AK020170.1 |
- |
1, 2 |
545 |
|
AK020448.1 |
- |
1, 2 |
317 |
|
AK045436.1 |
- |
1, 2 |
338 |
|
AK076594.1 |
- |
1, 2 |
350 |
|
BC046301.1 |
- |
1, 2 |
336 |
|
BC061099.1 |
- |
1, 2 |
329 |
6742251 |
BY719456.1 |
- |
1, 2 |
542 |
|
View Full Product Details