Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023058.3 | NP_075545.2 | 2, 3 | 802 | |
GenBank mRNA | AF175892.1 | - | 2, 3 | 686 | |
AK158550.1 | - | 2, 3 | 803 | ||
AK165164.1 | - | 2, 3 | 669 | ||
AK181294.1 | - | 1, 2 | 44 | ||
BC025061.1 | - | 2, 3 | 525 | 2609421 |