Gene Transcripts

Gene Symbol
Arpc1b
Entrez Gene ID
11867
Gene Name
actin related protein 2/3 complex, subunit 1B
Gene Aliases
41kDa, AA408064, AA408534, AA571392, AF007010, AW208418, L72, SOP2Hs, p41-ARC
Chromosome Location
Chr. 5: 145114256 - 145128186 on Build GRCm38
UniGene ID
Mm.30010
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023142.2 NP_075631.2 9 1130
GenBank mRNA AF162768.1 - 8 1055
AK002274.1 - 9 1131
AK005378.1 - 9 1131
AK147731.1 - 9 1098
AK148145.1 - 9 1124
AK151911.1 - 9 1130
AK153510.1 - 9 1130
AK157098.1 - 9 1121
AK159371.1 - 9 1129
AK159477.1 - 9 1020
AK167978.1 - 9 1060
AK168774.1 - 9 1128
AK170759.1 - 9 1120
AK171364.1 - 9 1092
AK172124.1 - 9 1098
BC003441.1 - 9 1062 2650641
BC010275.1 - 9 1072 3589768
BC020404.1 - 6 658
BC092051.1 - 9 1086 3668682

View Full Product Details