Gene Transcripts
- Gene Symbol
- Grina
- Entrez Gene ID
- 66168
- Gene Name
- glutamate receptor, ionotropic, N-methyl D-aspartate-associated protein 1 (glutamate binding)
- Gene Aliases
- 1110025J15Rik, Lag, Tmbim3
- Chromosome Location
- Chr. 15: 76246740 - 76249911 on Build GRCm38
- UniGene ID
- Mm.41665
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_023168.3 |
NP_075657.1
|
3, 4 |
524 |
|
GenBank mRNA |
AF182040.1 |
- |
3, 4 |
507 |
|
AK003920.1 |
- |
3, 4 |
533 |
|
AK179160.1 |
- |
2, 3 |
148 |
|
AK189399.1 |
- |
2, 3 |
148 |
|
AK200011.1 |
- |
2, 3 |
175 |
|
AK212726.1 |
- |
2, 3 |
191 |
|
BC010802.1 |
- |
3, 4 |
557 |
4236429 |
BC019157.1 |
- |
3, 4 |
545 |
|
BC037667.1 |
- |
3, 4 |
524 |
5376054 |
View Full Product Details