Gene Transcripts

Gene Symbol
Ndc80
Entrez Gene ID
67052
Gene Name
NDC80 homolog, kinetochore complex component (S. cerevisiae)
Gene Aliases
2610020P18Rik, AV139730, HEC, HEC1, Kntc2
Chromosome Location
Chr. 17: 71496100 - 71526857 on Build GRCm38
UniGene ID
Mm.225956
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023294.2 NP_075783.2 5 614
GenBank mRNA AF326730.1 - 5 627
AK011497.1 - 5 584
AK132008.1 - 6 603
AK136236.1 - 4 433
AK163317.1 - 5 565
AK169427.1 - 5 571
BC020131.1 - 5 550 3709641

View Full Product Details