Gene Transcripts

Gene Symbol
Gtf2a1l
Entrez Gene ID
71828
Gene Name
general transcription factor IIA, 1-like
Gene Aliases
1700011N16Rik, Alf, Gtf2a1lf
Chromosome Location
Chr. 17: 88665502 - 88715157 on Build GRCm38
UniGene ID
Mm.61330
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023630.2 NP_076119.2 2 155
GenBank mRNA AF250835.1 - 2 123
AK005881.1 - 2 130
BC050756.1 - 2 138 6741860
BY092414.1 - 2 155

View Full Product Details