Gene Transcripts

Gene Symbol
Aox4
Entrez Gene ID
71872
Gene Name
aldehyde oxidase 4
Gene Aliases
2310003G12Rik, Aoh2
Chromosome Location
Chr. 1: 58210397 - 58268600 on Build GRCm38
UniGene ID
Mm.244525
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023631.2 NP_076120.2 2, 3 290
GenBank mRNA AF233581.1 - 2, 3 217
AK158427.1 - 2, 3 291
AK160953.1 - 2, 3 256
BC117975.1 - 2, 3 217 40098789
BC117976.1 - 2, 3 217 40098791

View Full Product Details