Gene Transcripts
- Gene Symbol
- Dnaja3
- Entrez Gene ID
- 83945
- Gene Name
- DnaJ (Hsp40) homolog, subfamily A, member 3
- Gene Aliases
- 1200003J13Rik, 1810053A11Rik, C81173, Tid-1, Tid1l
- Chromosome Location
- Chr. 16: 4684070 - 4707695 on Build GRCm38
- UniGene ID
- Mm.248337
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135112.1 |
NP_001128584.1
|
1, 2 |
283 |
|
NM_023646.4 |
NP_076135.3
|
1, 2 |
283 |
|
GenBank mRNA |
AF325535.1 |
- |
1, 2 |
324 |
|
AF326358.1 |
- |
1, 2 |
324 |
|
AK004575.1 |
- |
1, 2 |
233 |
|
AK004910.1 |
- |
1, 2 |
235 |
|
AK007852.1 |
- |
1, 2 |
224 |
|
AK031250.1 |
- |
1, 2 |
240 |
|
AK167511.1 |
- |
1, 2 |
245 |
|
AK214348.1 |
- |
|
16 |
|
AY009320.1 |
- |
1, 2 |
324 |
|
BC003920.1 |
- |
1, 2 |
216 |
|
BC027240.1 |
- |
1, 2 |
245 |
2654866 |
CJ148764.1 |
- |
1, 2 |
284 |
|
View Full Product Details