Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001291080.1 | NP_001278009.1 | 3 | 438 | |
NM_024176.2 | NP_077138.1 | 3 | 438 | ||
GenBank mRNA | AK005445.1 | - | 2 | 173 | |
AK010152.1 | - | 3 | 392 | ||
AK087456.1 | - | 3 | 264 | ||
AK154251.1 | - | 2 | 218 | ||
BC002090.1 | - | 3 | 375 | 3488139 | |
CT010290.1 | - | 3 | 173 |