Gene Transcripts
- Gene Symbol
- Scyl1
- Entrez Gene ID
- 78891
- Gene Name
- SCY1-like 1 (S. cerevisiae)
- Gene Aliases
- 2810011O19Rik, C85140, Ntkl, mdf, mfd, p105
- Chromosome Location
- Chr. 19: 5758427 - 5771414 on Build GRCm38
- UniGene ID
- Mm.276063
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_023912.2 |
NP_076401.1
|
6, 7 |
1087 |
|
GenBank mRNA |
AF276514.1 |
- |
6, 7 |
1087 |
|
AK019240.1 |
- |
6, 7 |
1080 |
|
AK134356.1 |
- |
6, 7 |
1086 |
|
AK144134.1 |
- |
6, 7 |
1073 |
|
AK145869.1 |
- |
6, 7 |
1107 |
|
AK161813.1 |
- |
6, 7 |
1086 |
|
AK200223.1 |
- |
1, 2 |
78 |
|
AK219353.1 |
- |
1, 2 |
78 |
|
BC034519.1 |
- |
6, 7 |
1073 |
3672524 |
EF437375.1 |
- |
2, 3 |
266 |
|
HM856325.1 |
- |
6, 7 |
1087 |
|
HM856326.1 |
- |
6, 7 |
1087 |
|
View Full Product Details