Gene Transcripts

Gene Symbol
AGA
Entrez Gene ID
175
Gene Name
aspartylglucosaminidase
Gene Aliases
AGU, ASRG, GA
Chromosome Location
Chr. 4: 177430774 - 177442503 on Build GRCh38
UniGene ID
Hs.207776
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000027.3 NP_000018.2 2 365
NM_001171988.1 NP_001165459.1 2 365
NR_033655.1 2 365
GenBank mRNA AK303370.1 - 2 271
AK312982.1 - 2 299
BC012392.1 - 2 288 3859317
CR541715.1 - 2 237
CX869892.1 - 2 242
DA346002.1 - 2 365
DA905583.1 - 2 355
M64073.1 - 2 240
X55762.1 - 2 264

View Full Product Details