Gene Transcripts

Gene Symbol
Hsbp1
Entrez Gene ID
68196
Gene Name
heat shock factor binding protein 1
Gene Aliases
0610007A03Rik, AA408769, Hsp25
Chromosome Location
Chr. 8: 119344538 - 119348929 on Build GRCm38
UniGene ID
Mm.358714
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024219.1 NP_077181.1 4 951
GenBank mRNA AK050550.1 - 4 1001
AK078498.1 - 4 983
AK145940.1 - 4 961
BC002153.1 - 4 951 3485301

View Full Product Details