Gene Transcripts
- Gene Symbol
- Sun1
- Entrez Gene ID
- 77053
- Gene Name
- Sad1 and UNC84 domain containing 1
- Gene Aliases
- 4632417G13Rik, 5730434D03Rik, Unc84a, mKIAA0810
- Chromosome Location
- Chr. 5: 139200558 - 139249839 on Build GRCm38
- UniGene ID
- Mm.210845
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001256115.1 |
NP_001243044.1
|
3, 4 |
360 |
|
NM_001256116.1 |
NP_001243045.1
|
2, 3 |
297 |
|
NM_001256117.1 |
NP_001243046.1
|
3, 4 |
360 |
|
NM_001256118.1 |
NP_001243047.1
|
2, 3 |
297 |
|
NM_024451.2 |
NP_077771.1
|
3, 4 |
360 |
|
GenBank mRNA |
AK014585.1 |
- |
3, 4 |
358 |
|
AK036187.1 |
- |
3, 4 |
337 |
|
AK160281.1 |
- |
3, 4 |
318 |
|
AK161959.1 |
- |
3, 4 |
332 |
|
AK162533.1 |
- |
3, 4 |
331 |
|
AK167686.1 |
- |
3, 4 |
332 |
|
AK208681.1 |
- |
1, 2 |
29 |
|
AK210229.1 |
- |
1 |
18 |
|
AY682989.1 |
- |
2, 3 |
277 |
|
BC047928.1 |
- |
3, 4 |
327 |
5321879 |
BC048156.1 |
- |
3, 4 |
329 |
5698402 |
BQ551727.1 |
- |
2, 3 |
301 |
|
CK389844.1 |
- |
3, 4 |
360 |
|
HQ402597.1 |
- |
2, 3 |
277 |
|
View Full Product Details