Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025453.3 | NP_079729.1 | 1 | 155 | |
GenBank mRNA | AK007532.1 | - | 1 | 156 | |
AK017154.1 | - | 1 | 157 | ||
AK017195.1 | - | 1 | 156 | ||
AK017209.1 | - | 1 | 164 | ||
BC117736.2 | - | 1 | 147 | 40089849 |