Gene Transcripts

Gene Symbol
Rep15
Entrez Gene ID
66532
Gene Name
RAB15 effector protein
Gene Aliases
2210417D09Rik
Chromosome Location
Chr. 6: 147032537 - 147033518 on Build GRCm38
UniGene ID
Mm.45888
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025620.2 NP_079896.1 1 529
GenBank mRNA AK008892.1 - 1 530
AK008964.1 - 1 532
BC024344.1 - 1 480 4988964

View Full Product Details