Gene Transcripts

Gene Symbol
Lonp2
Entrez Gene ID
66887
Gene Name
lon peptidase 2, peroxisomal
Gene Aliases
1300002A08Rik, AU015403, Lonp
Chromosome Location
Chr. 8: 86624043 - 86716636 on Build GRCm38
UniGene ID
Mm.324550
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025827.3 NP_080103.1 4 818
GenBank mRNA AK004843.1 - 4 732
AK050231.1 - 4 727
AK088348.1 - 4 819
AK151103.1 - 4 764
AK168791.1 - 4 764
AK168956.1 - 4 764
AK169656.1 - 732
AK218948.1 - 1 83
BC049090.1 - 4 754 6401542

View Full Product Details