Gene Transcripts

Gene Symbol
Ift22
Entrez Gene ID
67286
Gene Name
intraflagellar transport 22
Gene Aliases
3110017O03Rik, AI835745, AI847427, Rabl5
Chromosome Location
Chr. 5: 136908150 - 136914509 on Build GRCm38
UniGene ID
Mm.41779
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026073.3 NP_080349.1 2, 3 206
GenBank mRNA AK005820.1 - 2, 3 166
AK160464.1 - 2, 3 217
BC009150.1 - 2, 3 206 2651447

View Full Product Details