Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026172.3 | NP_080448.1 | 1, 2 | 232 | |
GenBank mRNA | AK002756.1 | - | 1, 2 | 112 | |
AK004725.2 | - | 1, 2 | 99 | ||
AK010473.1 | - | 1, 2 | 93 | ||
AK015692.1 | - | 1, 2 | 103 | ||
AK146003.1 | - | 1, 2 | 103 | ||
AK168490.1 | - | 1, 2 | 100 | ||
BC046972.1 | - | 1, 2 | 232 | 6469277 | |
CT010405.1 | - | 1, 2 | 76 |