Gene Transcripts

Gene Symbol
Slc47a1
Entrez Gene ID
67473
Gene Name
solute carrier family 47, member 1
Gene Aliases
1300013J15Rik, AI036982, MATE1, mMATE1
Chromosome Location
Chr. 11: 61343400 - 61378781 on Build GRCm38
UniGene ID
Mm.100741
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026183.5 NP_080459.2 11 1015
GenBank mRNA AK004994.2 - 12 1371
AK009038.1 - 6 635
BC031436.1 - 11 999

View Full Product Details