Gene Transcripts

Gene Symbol
TYRO3
Entrez Gene ID
7301
Gene Name
TYRO3 protein tyrosine kinase
Gene Aliases
BYK, Dtk, Etk-2, RSE, Rek, Sky, Tif
Chromosome Location
Chr. 15: 41559022 - 41579338 on Build GRCh38
UniGene ID
Hs.381282
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006293.3 NP_006284.2 12 1759
XM_017022542.1 XP_016878031.1 12 1724
XM_017022543.1 XP_016878032.1 12 1560
GenBank mRNA AB209431.1 - 12 1788
AK295496.1 - 1 54
BC029925.2 - 4 409
BC034960.1 - 12 1541
BC049368.1 - 12 1533 4821587
BC051756.1 - 12 1538 6095596
D17517.1 - 12 1747
D50479.1 - 12 1519
U02566.1 - 9 970
U05682.1 - 12 1529

View Full Product Details