Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026560.4 | NP_080836.3 | 2 | 387 | |
GenBank mRNA | AK010905.1 | - | 2 | 379 | |
AK020070.1 | - | 2 | 343 | ||
AK031571.1 | - | 2 | 315 | ||
AK076065.1 | - | 2 | 344 | ||
AK076422.1 | - | 2 | 377 | ||
AK077717.1 | - | 2 | 340 | ||
AK077920.1 | - | 2 | 310 | ||
AK078404.1 | - | 2 | 340 | ||
AK082800.1 | - | 2 | 352 | ||
AY550907.1 | - | 2 | 377 | ||
BC068181.1 | - | 2 | 335 | 30634239 |