Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026499.4 | NP_080775.3 | 6 | 1030 | |
GenBank mRNA | AK004831.1 | - | 6 | 1011 | |
AK011561.1 | - | 1 | 36 | ||
AK012605.1 | - | 6 | 778 | ||
AK051608.1 | - | 4 | 1183 | ||
AK133872.1 | - | 6 | 1011 | ||
AK146293.1 | - | 6 | 1008 | ||
AK159519.1 | - | 6 | 1008 | ||
AK171709.1 | - | 5 | 2294 | ||
AK214648.1 | - | 1 | 51 | ||
BC012039.1 | - | 6 | 1017 | 2648111 | |
BC029312.1 | - | 1 | 124 |