Gene Transcripts

Gene Symbol
Ndufaf4
Entrez Gene ID
68493
Gene Name
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 4
Gene Aliases
1110007M04Rik, 3000003G13Rik, AW214064
Chromosome Location
Chr. 4: 24898083 - 24905001 on Build GRCm38
UniGene ID
Mm.204831
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026742.4 NP_081018.1 3 592
GenBank mRNA AK003553.2 - 3 547
AK034728.1 - 3 575
AK160341.1 - 3 588
BC028764.1 - 3 561 1195846

View Full Product Details