Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000187.3 | NP_000178.2 | 3 | 604 | |
XM_005247412.1 | XP_005247469.1 | 3 | 604 | ||
XM_005247413.1 | XP_005247470.1 | 3 | 604 | ||
XM_005247414.4 | XP_005247471.1 | 3 | 604 | ||
XM_011512746.1 | XP_011511048.1 | 3 | 604 | ||
XM_017006277.1 | XP_016861766.1 | 3 | 217 | ||
GenBank mRNA | AF045167.1 | - | 3 | 311 | |
AK290782.1 | - | 3 | 310 | ||
AK313563.1 | - | 3 | 588 | ||
BC020792.1 | - | 3 | 310 | 4716316 | |
BC071757.1 | - | 3 | 242 | 6714217 | |
U63008.1 | - | 3 | 273 | ||
Z75048.1 | - | 3 | 273 |