Gene Transcripts
- Gene Symbol
- Ptcd3
- Entrez Gene ID
- 69956
- Gene Name
- pentatricopeptide repeat domain 3
- Gene Aliases
- 2610034F17Rik, 2810422B04Rik, AA589622, AU045708, MRP-S39
- Chromosome Location
- Chr. 6: 71880638 - 71908772 on Build GRCm38
- UniGene ID
- Mm.30256
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_027275.3 |
NP_081551.2
|
18, 19 |
1467 |
|
GenBank mRNA |
AK013131.1 |
- |
4, 5 |
246 |
|
AK146005.1 |
- |
18, 19 |
1456 |
|
AK146172.1 |
- |
18, 19 |
1468 |
|
AK172480.1 |
- |
19, 20 |
1532 |
|
BC026759.1 |
- |
2, 3 |
122 |
|
BC055844.1 |
- |
4, 5 |
294 |
|
BC060687.1 |
- |
9, 10 |
688 |
|
BC084595.1 |
- |
9, 10 |
717 |
|
BC115434.2 |
- |
18, 19 |
1463 |
40094203 |
View Full Product Details