Gene Transcripts

Gene Symbol
Cmtm8
Entrez Gene ID
70031
Gene Name
CKLF-like MARVEL transmembrane domain containing 8
Gene Aliases
2700018N07Rik, AA408515, Cklfsf8
Chromosome Location
Chr. 9: 114789343 - 114844152 on Build GRCm38
UniGene ID
Mm.150064
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027294.2 NP_081570.1 3 635
GenBank mRNA AK012262.1 - 3 636
AY243513.1 - 3 670
BC027554.1 - 3 557 1382818

View Full Product Details