Gene Transcripts

Gene Symbol
Lrr1
Entrez Gene ID
69706
Gene Name
leucine rich repeat protein 1
Gene Aliases
2410005L11Rik, AU021780, LRR-1, Ppil5
Chromosome Location
Chr. 12: 69168814 - 69179010 on Build GRCm38
UniGene ID
Mm.28847
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK010419.1 - 2, 3 349
BC022648.1 - 2, 3 310
BC117053.1 - 2, 3 335
BC117055.1 - 2, 3 335

View Full Product Details