Gene Transcripts

Gene Symbol
Eqtn
Entrez Gene ID
67753
Gene Name
equatorin, sperm acrosome associated
Gene Aliases
1700028B15Rik, 4930579C15Rik, Afaf, C81058, Eqtl, Eqts
Chromosome Location
Chr. 4: 94875696 - 94929950 on Build GRCm38
UniGene ID
Mm.25097
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027089.4 NP_081365.2 8 814
GenBank mRNA AB438105.1 - 8 723
AK006446.1 - 8 765
BC089517.1 - 8 781 6740450
EU828781.1 - 8 799

View Full Product Details