Gene Transcripts

Gene Symbol
Rufy3
Entrez Gene ID
52822
Gene Name
RUN and FYVE domain containing 3
Gene Aliases
2810428M05Rik, 6330416M07Rik, AW455998, AW538594, D5Bwg0860e, Ripx, Rpipx
Chromosome Location
Chr. 5: 88564780 - 88651417 on Build GRCm38
UniGene ID
Mm.270469
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289774.1 NP_001276703.1 2 510
NM_001289775.1 NP_001276704.1 2 510
NM_001289776.1 NP_001276705.1 3 932
NM_001289777.1 NP_001276706.1 4 757
NM_027530.3 NP_081806.1 2 878
GenBank mRNA AK013185.1 - 4 758
AK018184.1 - 2 856
AK045610.1 - 2 812
AK083242.1 - 2 813
AK148795.1 - 2 174
AK154941.1 - 2 511
AK160820.1 - 2 511
BC017648.1 - 2 267 4527858
BC049127.1 - 2 817 6821567
BC058259.1 - 3 651 6469137
CT010404.1 - 2 251
DV068924.1 - 3 634

View Full Product Details