Gene Transcripts

Gene Symbol
Sik3
Entrez Gene ID
70661
Gene Name
SIK family kinase 3
Gene Aliases
5730525O22Rik, 9030204A07Rik, AI447252, AI846254, AI849445, Qsk, SIK-3, mKIAA0999
Chromosome Location
Chr. 9: 46012820 - 46224194 on Build GRCm38
UniGene ID
Mm.219459
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027498.3 NP_081774.3 5 752
GenBank mRNA AK017789.1 - 4 502
BC063268.1 - 5 548 4187769
BC080688.1 - 5 694 6515742

View Full Product Details